missing translation for 'onlineSavingsMsg'
Learn More

Potassium Acid Tartrate, Powder, FCC, 99-101%, Spectrum™ Chemical

Catalog No. 18605439
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605439 50 kg Poly Bottle
18605436 500 g Poly Bottle
18605437 2.5 kg Poly Bottle
18605438 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605439 Supplier Spectrum Chemical Mfg Cor Supplier No. P120650KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

P1206, 868-14-4, C4H5KO6

Spectrum™ Chemical Potassium Acid Tartrate, Powder, FCC or potassium bitartrate is called cream of tartar in the food industry. It is useful to stabilize egg whites and whipped cream, prevents sugar syrups from crystallizing and reduces discoloration of boiled vegetables. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 868-14-4
Molecular Formula C4H5KO6
Molecular Weight (g/mol) 188.18
MDL Number MFCD00065392,MFCD00071626,MFCD00064206
InChI Key KYKNRZGSIGMXFH-ZVGUSBNCSA-M
IUPAC Name potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
SMILES [K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 50 kg
Percent Purity 99 to 101%
Grade FCC
Packaging Poly Bottle
Chemical Name or Material Potassium Acid Tartrate, Powder