accessibility menu, dialog, popup

UserName

CAS RN 866405-64-3

NONNNN

CAS RN 866405-64-3

IUPAC Name: 4-(6-{4-[2-(piperidin-1-yl)ethoxy]phenyl}pyrazolo[1,5-a]pyrimidin-3-yl)pyridine
Synonyms: 4-(6-{4-[2-(piperidin-1-yl)ethoxy]phenyl}pyrazolo[1,5-a]pyrimidin-3-yl)pyridine
Molecular Weight (g/mol): 399.5
Molecular Formula: C24H25N5O
InChi Key: XHBVYDAKJHETMP-UHFFFAOYSA-N
SMILES: C(CN1CCCCC1)OC1=CC=C(C=C1)C1=CN2N=CC(=C2N=C1)C1=CC=NC=C1
Form 
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results