accessibility menu, dialog, popup

UserName

BrH3CNCH3CH3CH3

CAS RN 866-97-7

IUPAC Name: tetrapentylazanium bromide
Synonyms: tetrapentylammonium bromide
Molecular Weight (g/mol): 378.48
Molecular Formula: C20H44BrN
InChi Key: SPALIFXDWQTXKS-UHFFFAOYSA-M
SMILES: [Br-].CCCCC[N+](CCCCC)(CCCCC)CCCCC
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)