missing translation for 'onlineSavingsMsg'
Learn More

Tetraamylammonium Bromide 98.0+%, TCI America™

Catalog No. T14325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14325G 5 g
T143225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14325G Supplier TCI America Supplier No. T14325G

Chemical Identifiers

CAS 866-97-7
Molecular Formula C20H44BrN
Molecular Weight (g/mol) 378.48
MDL Number MFCD00011856
InChI Key SPALIFXDWQTXKS-UHFFFAOYSA-M
Synonym tetrapentylammonium bromide, tetraamylammonium bromide, tetra-n-amylammonium bromide, 1-pentanaminium, n,n,n-tripentyl-, bromide 1:1, tetrapentylazanium bromide, n,n,n-tripentylpentan-1-aminium bromide, tetraamylammoniumbromide, 1-pentanaminium, n,n,n-tripentyl-, bromide, acmc-209qb0, tetrapentyl ammonium bromide
PubChem CID 70086
IUPAC Name tetrapentylazanium bromide
SMILES [Br-].CCCCC[N+](CCCCC)(CCCCC)CCCCC

Specifications

Melting Point 101°C
Color White
Quantity 5 g
Formula Weight 378.48
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetraamylammonium Bromide
EINECSNumber (2)-0186
TSCA Yes