accessibility menu, dialog, popup

UserName

CAS RN 86595-37-1

CH3H3CCH3BOHHO(E)

CAS RN 86595-37-1

IUPAC Name: [(1E)-3,3-dimethylbut-1-en-1-yl]boronic acid
Synonyms: (1E)-3,3-dimethylbut-1-en-1-ylboronic acid
Molecular Weight (g/mol): 127.98
Molecular Formula: C6H13BO2
InChi Key: CTIWKIMYFQSVBZ-SNAWJCMRSA-N
SMILES: CC(C)(C)\C=C\B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results