missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butyl-trans-vinylboronic acid, 97%

Catalog No. AAH52627MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52627MD 250 mg
AAH5262703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52627MD Supplier Thermo Scientific Chemicals Supplier No. H52627MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 86595-37-1 | C6H13BO2 | 127.978 g/mol

2-tert-Butyl-trans-vinylboronic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-tert-Butyl-trans-vinylboronic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 86595-37-1
Molecular Formula C6H13BO2
Molecular Weight (g/mol) 127.978
MDL Number MFCD01074624
InChI Key CTIWKIMYFQSVBZ-SNAWJCMRSA-N
Synonym 3,3-dimethyl-1-butenylboronic acid, e-3,3-dimethylbut-1-en-1-yl boronic acid, 2-t-butyl-e-vinylboronic acid, 2-tert-butyl-e-vinylboronic acid, e-3,3-dimethylbut-1-enyl boronic acid, 1-butenyl-3,3-dimethyboronic acid, 1e-3,3-dimethylbut-1-en-1-ylboronic acid, boronic acid, b-3,3-dimethyl-1-buten-1-yl, 3,3-dimethylbut-1-enylboronic acid
PubChem CID 5706775
IUPAC Name [(E)-3,3-dimethylbut-1-enyl]boronic acid
SMILES B(C=CC(C)(C)C)(O)O

Specifications

Melting Point 78°C to 80°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 127.98
Percent Purity 97%
Chemical Name or Material 2-tert-Butyl-trans-vinylboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only