accessibility menu, dialog, popup

UserName

CAS RN 86303-23-3

IUPAC Name: N-[3-(4-{7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}-N-[3-(2,3,4,5,6-pentahydroxyhexanamido)propyl]pentanamido)propyl]-2,3,4,5,6-pentahydroxyhexanamide
Synonyms: deoxy-bigchap
Molecular Weight (g/mol): 862.07
Molecular Formula: C42H75N3O15
InChi Key: OJSUWTDDXLCUFR-UHFFFAOYNA-N
SMILES: CC(CCC(=O)N(CCCNC(=O)C(O)C(O)C(O)C(O)CO)CCCNC(=O)C(O)C(O)C(O)C(O)CO)C1CCC2C3CCC4CC(O)CCC4(C)C3CC(O)C12C
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results