missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ CHAP, Big, Deoxy, ≥95%, Calbiochem™,

Catalog No. 2564551GM
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2564551GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2564551GM Supplier MilliporeSigma™ Supplier No. 2564551GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 86303-23-3
Molecular Formula C42H75N3O15
Molecular Weight (g/mol) 862.068
InChI Key OJSUWTDDXLCUFR-HGZMBBKESA-N
Synonym N,N-bis-(3-D-Gluconamidopropyl)deoxycholamide
PubChem CID 446320
IUPAC Name (2R,3S,4R,5R)-N-[3-[[(4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]-[3-[[(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoyl]amino]propyl]amino
SMILES CC(CCC(=O)N(CCCNC(=O)C(C(C(C(CO)O)O)O)O)CCCNC(=O)C(C(C(C(CO)O)O)O)O)C1CCC2C1(C(CC3C2CCC4C3(CCC(C4)O)C)O)C

Specifications

Color White
Quantity 1 g
Packaging Glass Bottle
Percent Purity ≥95% (HPLC)