accessibility menu, dialog, popup

UserName

CAS RN 86156-93-6

IUPAC Name: 3-fluoro-4-nitrobenzene-1-sulfonyl chloride
Synonyms: 3-fluoro-4-nitrobenzenesulfonyl chloride
Molecular Weight (g/mol): 239.6
Molecular Formula: C6H3ClFNO4S
InChi Key: AAWFUSFNPBRQDE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(F)C=C(C=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results