missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-nitrobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog No. AAH3197903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3197903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3197903 Supplier Thermo Scientific Chemicals Supplier No. H3197903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 86156-93-6 | Molecular Formula: C6H3ClFNO4S | Molecular Weight: 239.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86156-93-6
Molecular Formula C6H3ClFNO4S
Molecular Weight (g/mol) 239.60
MDL Number MFCD03412240
InChI Key AAWFUSFNPBRQDE-UHFFFAOYSA-N
Synonym 3-fluoro-4-nitrobenzene-1-sulfonyl chloride, 3-fluoro-4-nitro-benzenesulfonyl chloride, 3-fluoro-4-nitrobenzenesulfonylchloride, 3-fluro-4-nitro-benzensulfonylchloride, 3-fluoro-4-nitrophenylsulfonyl chloride, benzenesulfonylchloride, 3-fluoro-4-nitro, benzenesulfonyl chloride, 3-fluoro-4-nitro, 3-fluoro-4-nitro-benzensulfonyl chloride, benzenesulfonyl chloride, 3-fluoro-4-nitro-9ci
PubChem CID 15569555
IUPAC Name 3-fluoro-4-nitrobenzenesulfonyl chloride
SMILES [O-][N+](=O)C1=C(F)C=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 44°C to 48°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 239.61
Percent Purity 98%
Chemical Name or Material 3-Fluoro-4-nitrobenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only