accessibility menu, dialog, popup

UserName

CAS RN 861306-04-9

H3COH2NOOH

CAS RN 861306-04-9

IUPAC Name: 3-amino-2-methoxybenzoic acid
Synonyms: 3-amino-2-methoxybenzoic acid
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: ZBQFWVIYNLHFMO-UHFFFAOYSA-N
SMILES: COC1=C(C=CC=C1N)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results