accessibility menu, dialog, popup

UserName

NOOOOFFFFF

CAS RN 86060-90-4

IUPAC Name: 1-(9H-fluoren-9-yl)methyl 2-(2,3,4,5,6-pentafluorophenyl) pyrrolidine-1,2-dicarboxylate
Synonyms: 1-(9H-fluoren-9-ylmethyl) 2-(2,3,4,5,6-pentafluorophenyl) pyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol): 503.43
Molecular Formula: C26H18F5NO4
InChi Key: CQBLOHXKGUNWRV-UHFFFAOYNA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2CCCN2C(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results