missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-proline pentafluorophenyl ester, 98%, Thermo Scientific™

Catalog No. AAH6345706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6345706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6345706 Supplier Thermo Scientific Chemicals Supplier No. H6345706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 86060-90-4
Molecular Formula C26H18F5NO4
Molecular Weight (g/mol) 503.425
MDL Number MFCD00065672
InChI Key CQBLOHXKGUNWRV-SFHVURJKSA-N
Synonym fmoc-pro-opfp, fmoc-l-pro-opfp, fmoc-l-proline pentafluorophenyl ester, 1,2-pyrrolidinedicarboxylicacid, 1-9h-fluoren-9-ylmethyl 2-pentafluorophenyl ester, 2s, n-9-fluorenylmethoxycarbonyl-l-proline pentafluorophenyl ester, 1-9h-fluorene-9-ylmethoxycarbonyl-l-proline pentafluorophenyl ester, 1-9h-fluoren-9-ylmethyl 2-2,3,4,5,6-pentafluorophenyl 2s-pyrrolidine-1,2-dicarboxylate, 1-o-9h-fluoren-9-ylmethyl 2-o-2,3,4,5,6-pentafluorophenyl 2s-pyrrolidine-1,2-dicarboxylate
PubChem CID 2733412
IUPAC Name 1-O-(9H-fluoren-9-ylmethyl) 2-O-(2,3,4,5,6-pentafluorophenyl) (2S)-pyrrolidine-1,2-dicarboxylate
SMILES C1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)OC5=C(C(=C(C(=C5F)F)F)F)F

Specifications

Melting Point 127°C to 129°C
Beilstein 4731096
Quantity 5 g
Formula Weight 503.42
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Fmoc-L-proline pentafluorophenyl ester
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold