accessibility menu, dialog, popup

UserName

CAS RN 86-86-2

NH2O

CAS RN 86-86-2

IUPAC Name: 2-(naphthalen-1-yl)acetamide
Synonyms: 1-naphthaleneacetamide
Molecular Weight (g/mol): 185.23
Molecular Formula: C12H11NO
InChi Key: XFNJVKMNNVCYEK-UHFFFAOYSA-N
SMILES: NC(=O)CC1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)