missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ 1-Naphthylacetamide, MilliporeSigma™ Supelco™

Catalog No. 111001853
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001853 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001853 Supplier Merck Emd Millipore Supplier No. 36732250MG
Agriculture

Specifications

Product Line PESTANAL
CAS 86-86-2
Physical Form Neat
Linear Formula C10H7 CH2 CONH2
MDL Number MFCD00004047
Melting Point 180°C to 183°C
Synonym 1-Naphthaleneacetamide
Type Analytical Standard
Quantity 250 mg
Formula Weight 185.22
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH