accessibility menu, dialog, popup

UserName

CAS RN 86-55-5

IUPAC Name: naphthalene-1-carboxylic acid
Synonyms: naphthoic acid calcium salt 1-naphthoic acid sodium salt 1-naphthoic acid 1-napthoic acid
Molecular Weight (g/mol): 172.18
Molecular Formula: C11H8O2
InChi Key: LNETULKMXZVUST-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)