missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthoic Acid 98.0+%, TCI America™

Catalog No. N002425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N002425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N002425G Supplier TCI America Supplier No. N002425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 86-55-5
Melting Point 164°C
Molecular Formula C11H8O2
Quantity 25 g
MDL Number MFCD00004007
Synonym 1-naphthoic acid, 1-naphthalenecarboxylic acid, 1-carboxynaphthalene, naphthoic acid, alpha-naphthoic acid, naphthalenecarboxylic acid, carboxynaphthalene, alpha-naphthylcarboxylic acid, naphthalene-alpha-carboxylic acid, unii-2niv4o66bh
InChI Key LNETULKMXZVUST-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=CC=C2C(=O)O
IUPAC Name naphthalene-1-carboxylic acid
Molecular Weight (g/mol) 172.183
PubChem CID 6847
ChEBI CHEBI:36466
Formula Weight 172.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Naphthoic Acid
Show More Show Less
EINECSNumber (4)-0397
RTECSNumber QL0960000
TSCA Yes