accessibility menu, dialog, popup

UserName

NClBrNH2

CAS RN 857730-21-3

IUPAC Name: 5-bromo-2-chloropyridin-4-amine
Synonyms: 5-bromo-2-chloropyridin-4-amine
Molecular Weight (g/mol): 207.46
Molecular Formula: C5H4BrClN2
InChi Key: MGZWZNBANVSZLM-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=NC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results