missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-5-bromo-2-chloropyridine, 95%, Thermo Scientific Chemicals

Catalog No. AAH2767403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2767403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2767403 Supplier Thermo Scientific Chemicals Supplier No. H2767403

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 857730-21-3
Molecular Formula C5H4BrClN2
Molecular Weight (g/mol) 207.455
MDL Number MFCD09037459
InChI Key MGZWZNBANVSZLM-UHFFFAOYSA-N
Synonym 4-amino-5-bromo-2-chloropyridine, 5-bromo-2-chloro-4-pyridinamine, 2-chloro-5-bromo-4-aminopyridine, 4-pyridinamine, 5-bromo-2-chloro, 5-bromo-2-chloro-pyridin-4-ylamine, 2-chloro-5-bromo-pyridine-4-ylamine, pubchem19507, acmc-209uo6, ksc447s7p, 2-chloro-4-amino-5-bromopyridine
PubChem CID 37030051
IUPAC Name 5-bromo-2-chloropyridin-4-amine
SMILES C1=C(C(=CN=C1Cl)Br)N

Specifications

Melting Point 127°C to 129°C
Quantity 1 g
Formula Weight 207.46
Percent Purity 95%
Chemical Name or Material 4-Amino-5-bromo-2-chloropyridine
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only