CAS RN 856866-72-3
IUPAC Name:
3-{3-fluoro-4-[6-(2-methyl-2H-1,2,3,4-tetrazol-5-yl)pyridin-3-yl]phenyl}-5-(hydroxymethyl)-1,3-oxazolidin-2-one
Synonyms:
3-{3-fluoro-4-[6-(2-methyl-1,2,3,4-tetrazol-5-yl)pyridin-3-yl]phenyl}-5-(hydroxymethyl)-1,3-oxazolidin-2-one
Molecular Weight (g/mol):
370.34
Molecular Formula:
C17H15FN6O3
InChi Key:
XFALPSLJIHVRKE-UHFFFAOYNA-N
SMILES:
CN1N=NC(=N1)C1=NC=C(C=C1)C1=C(F)C=C(C=C1)N1CC(CO)OC1=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals Tedizolid
CAS: 856866-72-3 Molecular Formula: C17H15FN6O3