missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tedizolid

Catalog No. AC467580010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467580010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC467580010 Supplier Thermo Scientific Chemicals Supplier No. 467580010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 856866-72-3 | C17H15FN6O3, C17H15FN6O3 | 370.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 856866-72-3
Molecular Formula C17H15FN6O3

Specifications

Quantity 1 g
Melting Point 201.0°C
Formula Weight 370.34
Chemical Name or Material Tedizolid

RUO – Research Use Only