accessibility menu, dialog, popup

UserName

CAS RN 85652-50-2

IUPAC Name: 30,34-bis(octyloxy)nonacyclo[18.10.2.2²,⁵.0³,¹⁶.0⁴,¹³.0⁶,¹¹.0¹⁷,³¹.0²²,²⁷.0²⁸,³²]tetratriaconta-1(30),2(34),3,5(33),6,8,10,13,15,17,19,22,24,26,28,31-hexadecaene-12,21-dione
Synonyms: 30,34-bis(octyloxy)nonacyclo[18.10.2.2²,⁵.0³,¹⁶.0⁴,¹³.0⁶,¹¹.0¹⁷,³¹.0²²,²⁷.0²⁸,³²]tetratriaconta-1(30),2(34),3,5(33),6,8,10,13,15,17,19,22,24,26,28,31-hexadecaene-12,21-dione
Molecular Weight (g/mol): 712.93
Molecular Formula: C50H48O4
InChi Key: LLPQZABTDLOYAL-UHFFFAOYSA-N
SMILES: CCCCCCCCOC1=C2C3=C(OCCCCCCCC)C=C4C5=CC=CC=C5C(=O)C5=CC=C(C6=CC=C7C(=O)C8=CC=CC=C8C(=C1)C7=C26)C3=C45
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)