missing translation for 'onlineSavingsMsg'
Learn More

Violanthrone-79, 98%

Catalog No. AAH5961603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5961603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5961603 Supplier Thermo Scientific Chemicals Supplier No. H5961603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 85652-50-2 | C50H48O4 | 712.93 g/mol

Violanthrone 79 is an n-channel organic semiconductor used for interface control of conventional n-type meta diode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Violanthrone 79 is an n-channel organic semiconductor used for interface control of conventional n-type meta diode.

Solubility
Soluble in chloroform.

Notes
Refrigerated. Incompatible with strong oxi

Chemical Identifiers

CAS 85652-50-2
Molecular Formula C50H48O4
Molecular Weight (g/mol) 712.93
MDL Number MFCD00491570
InChI Key LLPQZABTDLOYAL-UHFFFAOYSA-N
Synonym violanthrone-79, violanthrone 79, anthra 9,1,2-cde benzo rst pentaphene-5,10-dione,16,17-bis octyloxy, anthra 9,1,2-cde benzo rst pentaphene-5,10-dione,1, 16,17-bis n-octyloxy anthra 9,1,2-cde benzo rst pentaphene-5,10-dione, 16,17-bis octyloxy-anthra 9,1,2-cde benzo rst pentaphene-5,10-dione, 16,17-bis octyloxy anthra 9,1,2-cde-benzo rst pentaphene-5,10-dione, 16,17-bis octyloxy anthra 9,1,2-cde benzo rst pentaphene-5,10-dione
PubChem CID 16217791
SMILES CCCCCCCCOC1=C2C3=C(C=CC4=C3C(=C1)C5=CC=CC=C5C4=O)C6=C7C2=C(C=C8C7=C(C=C6)C(=O)C9=CC=CC=C98)OCCCCCCCC

Specifications

Melting Point 193°C to 196°C
Quantity 1 g
Solubility Information Soluble in chloroform.
Formula Weight 712.91
Percent Purity 98%
Chemical Name or Material Violanthrone-79
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only