CAS RN 85553-55-5
IUPAC Name:
[3-(pyridin-4-yl)phenyl]methanol
Synonyms:
[3-(pyridin-4-yl)phenyl]methanol
Molecular Weight (g/mol):
185.23
Molecular Formula:
C12H11NO
InChi Key:
ZORHPGKUTIFODF-UHFFFAOYSA-N
SMILES:
OCC1=CC=CC(=C1)C1=CC=NC=C1
1
–
2
of
2
results
2
(3-Pyrid-4-ylphenyl)methanol, 97%, Thermo Scientific™
CAS: 85553-55-5 Molecular Formula: C12H11NO Molecular Weight (g/mol): 185.23 MDL Number: MFCD06659083 InChI Key: ZORHPGKUTIFODF-UHFFFAOYSA-N PubChem CID: 2795564 IUPAC Name: (3-pyridin-4-ylphenyl)methanol SMILES: OCC1=CC=CC(=C1)C1=CC=NC=C1