accessibility menu, dialog, popup

UserName

H3CNCH3NSOOFSOOF

CAS RN 852620-97-4

IUPAC Name: 1-methyl-1-propylpyrrolidin-1-ium; bis(fluorosulfonyl)azanide
Synonyms: 1-methyl-1-propylpyrrolidin-1-ium; bis(fluorosulfonyl)azanide
Molecular Weight (g/mol): 308.36
Molecular Formula: C8H18F2N2O4S2
InChi Key: RXKLBLXXQQRGJH-UHFFFAOYSA-N
SMILES: CCC[N+]1(C)CCCC1.FS(=O)(=O)[N-]S(F)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results