missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1-propylpyrrolidinium Bis(fluorosulfonyl)imide 98.0+%, TCI America™

Catalog No. M29805G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M29805G Supplier TCI America Supplier No. M29805G

Chemical Identifiers

CAS 852620-97-4
Molecular Formula C8H18F2N2O4S2
Molecular Weight (g/mol) 308.359
MDL Number MFCD17018948
InChI Key RXKLBLXXQQRGJH-UHFFFAOYSA-N
PubChem CID 121234258
IUPAC Name bis(fluorosulfonyl)azanide;1-methyl-1-propylpyrrolidin-1-ium
SMILES CCC[N+]1(CCCC1)C.[N-](S(=O)(=O)F)S(=O)(=O)F

Specifications

Melting Point -9°C
Color Yellow
Quantity 5 g
Formula Weight 308.36
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Methyl-1-propylpyrrolidinium Bis(fluorosulfonyl)imide