CAS RN 85261-20-7
IUPAC Name:
N-methyl-N-[(2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl]decanamide
Synonyms:
N-methyl-N-[(2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl]decanamide
Molecular Weight (g/mol):
349.47
Molecular Formula:
C17H35NO6
InChi Key:
UMWKZHPREXJQGR-XOSAIJSUSA-N
SMILES:
CCCCCCCCCC(=O)N(C)C[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO
1
–
1
of
1
results
1
N-Decanoyl-N-methyl-D-glucamine
CAS: 85261-20-7 Molecular Formula: C17H35NO6 Molecular Weight (g/mol): 349.468 MDL Number: MFCD00036801 InChI Key: UMWKZHPREXJQGR-XOSAIJSUSA-N Synonym: n-oxodecyl meglumine,unii-9q27za5m8g,n-methyl-n-2s,3r,4r,5r-2,3,4,5,6-pentahydroxyhexyl decanamide,decanoyl n-methylglucamide,decanoylmethylglucamine,d-glucitol, 1-deoxy-1-methyl 1-oxodecyl amino,d-glucitol, 1-deoxy-1-methyl 1-oxodecyl amino-, stereoisomer PubChem CID: 174432 IUPAC Name: N-methyl-N-[(2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl]decanamide SMILES: CCCCCCCCCC(=O)N(C)CC(C(C(C(CO)O)O)O)O