missing translation for 'onlineSavingsMsg'
Learn More

N-Decanoyl-N-methyl-D-glucamine

Catalog No. AAJ6017306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6017306 5 g
AAJ6017303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6017306 Supplier Thermo Scientific Chemicals Supplier No. J6017306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Non-ionic detergent for solubilizing membrane proteins | CAS: 85261-20-7 | C17H35NO6 | 349.468 g/mol

Non-ionic detergent for solubilizing membrane proteins

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 85261-20-7
Molecular Formula C17H35NO6
Molecular Weight (g/mol) 349.468
MDL Number MFCD00036801
InChI Key UMWKZHPREXJQGR-XOSAIJSUSA-N
Synonym n-oxodecyl meglumine, unii-9q27za5m8g, n-methyl-n-2s,3r,4r,5r-2,3,4,5,6-pentahydroxyhexyl decanamide, decanoyl n-methylglucamide, decanoylmethylglucamine, d-glucitol, 1-deoxy-1-methyl 1-oxodecyl amino, d-glucitol, 1-deoxy-1-methyl 1-oxodecyl amino-, stereoisomer
PubChem CID 174432
IUPAC Name N-methyl-N-[(2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl]decanamide
SMILES CCCCCCCCCC(=O)N(C)CC(C(C(C(CO)O)O)O)O

Specifications

Melting Point 91°C to 93°C
Color White
Quantity 5 g
Beilstein 6974359
Solubility Information Soluble in methanol at 50mg/ml; Also soluble in water at 10mg/ml 20 °C
Formula Weight 349.46
Physical Form Powder
Chemical Name or Material N-Decanoyl-N-methyl-D-glucamine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only