accessibility menu, dialog, popup

UserName

CAS RN 852180-72-4

CH3NHN

CAS RN 852180-72-4

IUPAC Name: methyl({[3-(pyridin-3-yl)phenyl]methyl})amine
Synonyms: methyl({[3-(pyridin-3-yl)phenyl]methyl})amine
Molecular Weight (g/mol): 198.27
Molecular Formula: C13H14N2
InChi Key: LZJRRTLTBULOHU-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC(=C1)C1=CN=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results