missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-(3-pyridin-3-ylbenzyl)amine, 97%, Thermo Scientific™

Catalog No. CC39846CB
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC39846CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC39846CB Supplier Thermo Scientific Chemicals Supplier No. CC39846CB

Chemical Identifiers

CAS 852180-72-4
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.269
MDL Number MFCD07368550
InChI Key LZJRRTLTBULOHU-UHFFFAOYSA-N
Synonym n-methyl-n-3-pyridin-3-ylbenzyl amine, methyl 3-pyridin-3-yl phenyl methyl amine, n-methyl-n-3-pyridin-3-yl benzylamine, benzenemethanamine,n-methyl-3-3-pyridinyl, n-methyl-1-3-pyridin-3-yl phenyl methanamine, n-methyl-1-3-pyridin-3-ylphenyl methanamine, n-methyl-3-pyridin-3-yl benzylamine, methyl 3-3-pyridyl phenyl methyl amine, 3-3-methylamino methyl phenyl pyridine
PubChem CID 7060576
IUPAC Name N-methyl-1-(3-pyridin-3-ylphenyl)methanamine
SMILES CNCC1=CC=CC(=C1)C2=CN=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 198.27
Percent Purity 97%
Chemical Name or Material N-Methyl-N-(3-pyridin-3-ylbenzyl)amine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.