accessibility menu, dialog, popup

UserName

CAS RN 852062-17-0

IUPAC Name: 3-fluoro-5-(trifluoromethyl)pyridin-2-amine
Synonyms: 3-fluoro-5-(trifluoromethyl)pyridin-2-amine
Molecular Weight (g/mol): 180.11
Molecular Formula: C6H4F4N2
InChi Key: HAVVQRHBCUBQGG-UHFFFAOYSA-N
SMILES: NC1=C(F)C=C(C=N1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results