missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-fluoro-5-(trifluoromethyl)pyridine, 97%

Catalog No. AAH3253206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3253206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3253206 Supplier Thermo Scientific Chemicals Supplier No. H3253206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 852062-17-0 | C6H4F4N2 | 180.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 852062-17-0
Molecular Formula C6H4F4N2
Molecular Weight (g/mol) 180.106
MDL Number MFCD10699117
InChI Key HAVVQRHBCUBQGG-UHFFFAOYSA-N
Synonym 2-amino-3-fluoro-5-trifluoromethyl pyridine, 3-fluoro-5-trifluoromethyl pyridin-2-amine, 2-amino-3-fluoro-5-trifluoromethylpyridine, 3-fluoro-5-trifluoromethyl-2-pyridylamine, 3-fluoro-5-trifluoromethyl pyridine-2-amine, 3-fluoro-5-trifluoromethyl-2-pyridinamine
PubChem CID 25067349
IUPAC Name 3-fluoro-5-(trifluoromethyl)pyridin-2-amine
SMILES C1=C(C=NC(=C1F)N)C(F)(F)F

Specifications

Melting Point 64°C to 68°C
Quantity 5 g
UN Number UN2811
Formula Weight 180.1
Percent Purity 97%
Chemical Name or Material 2-Amino-3-fluoro-5-(trifluoromethyl)pyridine
Hazard Category H301+H311-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only