CAS RN 851883-08-4
IUPAC Name:
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,2-diamine
Synonyms:
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,2-diamine
Molecular Weight (g/mol):
234.11
Molecular Formula:
C12H19BN2O2
InChi Key:
SVUDHDDKOKWBNK-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=CC(N)=C(N)C=C1
1
–
1
of
1
results
1
Promotions Available
3,4-Diaminophenylboronic acid pinacol ester, 97%
3,4-Diaminophenylboronic acid pinacol ester, 97%
CAS: 851883-08-4 Molecular Formula: C12H19BN2O2 Molecular Weight (g/mol): 234.11 MDL Number: MFCD09027073 InChI Key: SVUDHDDKOKWBNK-UHFFFAOYSA-N Synonym: 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene-1,2-diamine,3,4-diaminophenylboronic acid, pinacol ester,3,4-diaminophenylboronic acid pinacol ester,pubchem22338,4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzene-1,2-diamine,4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2-benzenediamine,4-tetramethyl-1,3,2-dioxaborolan-2-yl benzene-1,2-diamine PubChem CID: 17750242 IUPAC Name: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,2-diamine SMILES: CC1(C)OB(OC1(C)C)C1=CC(N)=C(N)C=C1