missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminophenylboronic acid pinacol ester, 97%

Catalog No. AC444760010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC444760010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC444760010 Supplier Thermo Scientific Chemicals Supplier No. 444760010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 851883-08-4 | C12H19BN2O2 | 234.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 851883-08-4
Molecular Formula C12H19BN2O2
Molecular Weight (g/mol) 234.11
MDL Number MFCD09027073
InChI Key SVUDHDDKOKWBNK-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene-1,2-diamine, 3,4-diaminophenylboronic acid, pinacol ester, 3,4-diaminophenylboronic acid pinacol ester, pubchem22338, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzene-1,2-diamine, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2-benzenediamine, 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzene-1,2-diamine
PubChem CID 17750242
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,2-diamine
SMILES CC1(C)OB(OC1(C)C)C1=CC(N)=C(N)C=C1

Specifications

Melting Point 95°C to 97°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 234.11
Percent Purity 97%
Chemical Name or Material 3, 4-Diaminophenylboronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only