CAS RN 85098-70-0
CAS RN 85098-70-0
IUPAC Name:
1-[2-(4-methoxyphenyl)ethyl]piperidin-4-amine
Synonyms:
1-[2-(4-methoxyphenyl)ethyl]piperidin-4-amine
Molecular Weight (g/mol):
234.34
Molecular Formula:
C14H22N2O
InChi Key:
HTGMIBQQTLUEKV-UHFFFAOYSA-N
SMILES:
COC1=CC=C(CCN2CCC(N)CC2)C=C1
1
–
1
of
1
results
1
4-Amino-1-[2-(4-methoxyphenyl)ethyl]piperidine 98.0+%, TCI America™
CAS: 85098-70-0 Molecular Formula: C14H22N2O Molecular Weight (g/mol): 234.34 MDL Number: MFCD01631147 InChI Key: HTGMIBQQTLUEKV-UHFFFAOYSA-N Synonym: 4-Amino-1-(4-methoxyphenethyl)piperidine, 1-(4-Methoxyphenethyl)-4-aminopiperidine, 1-[2-(4-Methoxyphenyl)ethyl]-4-aminopiperidine PubChem CID: 3020332 IUPAC Name: 1-[2-(4-methoxyphenyl)ethyl]piperidin-4-amine SMILES: COC1=CC=C(CCN2CCC(N)CC2)C=C1