accessibility menu, dialog, popup

UserName

ONHNNN(Z)(E/Z)

CAS RN 85-86-9

IUPAC Name: (1Z)-1-{2-[4-(2-phenyldiazen-1-yl)phenyl]hydrazin-1-ylidene}-1,2-dihydronaphthalen-2-one
Synonyms: sudan III
Molecular Weight (g/mol): 352.4
Molecular Formula: C22H16N4O
InChi Key: HTPQPMPFXUWUOT-SRURNFRUSA-N
SMILES: O=C1C=CC2=CC=CC=C2\C1=N\NC1=CC=C(C=C1)N=NC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
Grade 
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)