missing translation for 'onlineSavingsMsg'
Learn More

G-Biosciences Sudan III Solution

Catalog No. S000787
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000787 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000787 Supplier G-Biosciences Supplier No. 7862203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Effectively visualize and identify lipid in tissue specimens with this red-orange histological stain

It dissolves in fats and oils, and when it binds to lipids, it stains them a distinctive red to orange color. This staining effect allows for easy visualization of fat content under a microscope.

Specifications

CAS 85-86-9
Molecular Weight (g/mol) 352.4 g/mol
Grade Research
Color Red
Quantity 250 mL