missing translation for 'onlineSavingsMsg'
Learn More

Sudan III 90.0+%, TCI America™

Catalog No. S014225G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S014225G 25 g
S0142100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S014225G Supplier TCI America Supplier No. S014225G
Only null left

Chemical Identifiers

CAS 85-86-9
Molecular Formula C22H16N4O
Molecular Weight (g/mol) 352.397
MDL Number MFCD00003905
InChI Key HTPQPMPFXUWUOT-UHFFFAOYSA-N
Synonym sudan iii, c.i. solvent red 23, 2-naphthalenol, 1-4-phenylazo phenyl azo, fhninjwbtrxebc-qszpnpogsa-n, sudaniii, sudan iiisolvent red 23, 2-naphthalenol, 1-2-4-2-phenyldiazenyl phenyl diazenyl, 1-2-4-2-phenyldiazen-1-yl phenyl diazen-1-yl naphthalen-2-ol, 1-e-4-e-phenylazo phenyl azo naphthalen-2-ol, 1-4-phenyldiazenylphenyl hydrazinylidene naphthalen-2-one
PubChem CID 6789251
IUPAC Name 1-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalen-2-one
SMILES C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC=CC=C43

Specifications

Melting Point 202°C
Quantity 25 g
Formula Weight 352.40
Percent Purity ≥90.0% (E)
Physical Form Crystalline Powder
Chemical Name or Material Sudan III
EINECSNumber (5)-3087
RTECSNumber QK4250000
TSCA Yes