missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sudan III, ≥96% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 85-86-9 |
| Physical Form | Neat |
| Linear Formula | C22H16N4 O |
| MDL Number | MFCD00003905 |
| Melting Point | 199°C |
| Synonym | 1-[4-(Phenylazo)phenylazo]-2-naphthol; Cerasin Red; Fat Ponceau G; Fat Soluble Sudan; Scarlet B; Solvent Red 23; Sudan G; Sudan Red BK; Tony Red |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 352.39 |
| Percent Purity | ≥96% (HPLC) |
| Show More |