accessibility menu, dialog, popup

UserName

FNClN

CAS RN 84946-20-3

IUPAC Name: 2-chloro-1-[(4-fluorophenyl)methyl]-1H-1,3-benzodiazole
Synonyms: 2-chloro-1-[(4-fluorophenyl)methyl]-1,3-benzodiazole
Molecular Weight (g/mol): 260.7
Molecular Formula: C14H10ClFN2
InChi Key: PGXALMVNIRPELS-UHFFFAOYSA-N
SMILES: FC1=CC=C(CN2C(Cl)=NC3=CC=CC=C23)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results