missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-(4-fluorobenzyl)benzimidazole 98.0+%, TCI America™

Catalog No. C28505G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28505G 5 g
C285025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28505G Supplier TCI America Supplier No. C28505G
Only null left

Chemical Identifiers

CAS 84946-20-3
Molecular Formula C14H10ClFN2
Molecular Weight (g/mol) 260.696
MDL Number MFCD00800240
InChI Key PGXALMVNIRPELS-UHFFFAOYSA-N
PubChem CID 3020241
IUPAC Name 2-chloro-1-[(4-fluorophenyl)methyl]benzimidazole
SMILES C1=CC=C2C(=C1)N=C(N2CC3=CC=C(C=C3)F)Cl

Specifications

Melting Point 83°C
Color White-Yellow
Quantity 5 g
Formula Weight 260.70
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-1-(4-fluorobenzyl)benzimidazole