CAS RN 84807-09-0
IUPAC Name:
4-(piperazin-1-yl)-1H-indole
Synonyms:
4-(piperazin-1-yl)-1H-indole
Molecular Weight (g/mol):
201.27
Molecular Formula:
C12H15N3
InChi Key:
YZKSXUIOKWQABW-UHFFFAOYSA-N
SMILES:
C1CN(CCN1)C1=C2C=CNC2=CC=C1
1
–
1
of
1
results
1
Promotions Available
4-(1-Piperazinyl)indole, 95%
4-(1-Piperazinyl)indole, 95%
CAS: 84807-09-0 Molecular Formula: C12H15N3 Molecular Weight (g/mol): 201.273 MDL Number: MFCD00831776 InChI Key: YZKSXUIOKWQABW-UHFFFAOYSA-N Synonym: 4-1-piperazinyl-1h-indole,4-piperazin-1-yl-1h-indole,4-piperazino-1h-indole,1h-indole, 4-1-piperazinyl,4-piperazino indole,1-4-indolyl piperazine,1h-indole-4-1-piperazinyl dihydrochloride,4-piperazinoindole,4-piperazino-indole,pubchem22668 PubChem CID: 594590 IUPAC Name: 4-piperazin-1-yl-1H-indole SMILES: C1CN(CCN1)C2=CC=CC3=C2C=CN3