accessibility menu, dialog, popup

UserName

ClONHOHNCl

CAS RN 84632-65-5

IUPAC Name: 3,6-bis(4-chlorophenyl)-1H,2H,4H,5H-pyrrolo[3,4-c]pyrrole-1,4-dione
Synonyms: 3,6-bis(4-chlorophenyl)-2H,5H-pyrrolo[3,4-c]pyrrole-1,4-dione
Molecular Weight (g/mol): 357.19
Molecular Formula: C18H10Cl2N2O2
InChi Key: JNNHVXMCVRYTTN-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C1=C2C(=O)NC(=C2C(=O)N1)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results