accessibility menu, dialog, popup

UserName

Br(?)

CAS RN 845457-53-6

IUPAC Name: 9-bromo-10-(phenanthren-9-yl)anthracene
Synonyms: 9-bromo-10-(phenanthren-9-yl)anthracene
Molecular Weight (g/mol): 433.35
Molecular Formula: C28H17Br
InChi Key: OBIXPXSRUDGFLE-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=CC2=C(C2=CC=CC=C12)C1=CC2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results