accessibility menu, dialog, popup

UserName

CAS RN 84539-22-0

IUPAC Name: 4-(5-bromopyrimidin-2-yl)morpholine
Synonyms: 4-(5-bromopyrimidin-2-yl)morpholine
Molecular Weight (g/mol): 244.09
Molecular Formula: C8H10BrN3O
InChi Key: CEXBOCXKAGPRHD-UHFFFAOYSA-N
SMILES: BrC1=CN=C(N=C1)N1CCOCC1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results