accessibility menu, dialog, popup

UserName

CAS RN 84466-85-3

CH3NN

CAS RN 84466-85-3

IUPAC Name: 1-[(1-methylpyrrolidin-2-yl)methyl]piperidine
Synonyms: 1-[(1-methylpyrrolidin-2-yl)methyl]piperidine
Molecular Weight (g/mol): 182.31
Molecular Formula: C11H22N2
InChi Key: VCVPSRADUBPOKJ-UHFFFAOYNA-N
SMILES: CN1CCCC1CN1CCCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results