accessibility menu, dialog, popup

UserName

CAS RN 84459-33-6

IUPAC Name: 2-bromo-4-chlorobenzaldehyde
Synonyms: 2-bromo-4-chlorobenzaldehyde
Molecular Weight (g/mol): 219.46
Molecular Formula: C7H4BrClO
InChi Key: AJOAHIKYBSZIEV-UHFFFAOYSA-N
SMILES: ClC1=CC(Br)=C(C=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results