missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-chlorobenzaldehyde 97.0+%, TCI America™

Catalog No. B55245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55245G 5 g
B552425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55245G Supplier TCI America Supplier No. B55245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 84459-33-6
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.46
MDL Number MFCD09056792
InChI Key AJOAHIKYBSZIEV-UHFFFAOYSA-N
PubChem CID 13154710
IUPAC Name 2-bromo-4-chlorobenzaldehyde
SMILES ClC1=CC(Br)=C(C=O)C=C1

Specifications

Melting Point 71°C
Color White-Yellow
Quantity 5 g
Formula Weight 219.46
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-chlorobenzaldehyde