accessibility menu, dialog, popup

UserName

CH3OOH

CAS RN 84-85-5

IUPAC Name: 4-methoxynaphthalen-1-ol
Synonyms: 4-methoxynaphthalen-1-ol
Molecular Weight (g/mol): 174.2
Molecular Formula: C11H10O2
InChi Key: BOTGCZBEERTTDQ-UHFFFAOYSA-N
SMILES: COC1=C2C=CC=CC2=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results