missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-1-naphthol 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 130°C |
| Quantity | 5 g |
| Formula Weight | 174.20 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | 4-Methoxy-1-naphthol |
Sécurité et manipulation
| EINECSNumber | (4)-0359 |
| TSCA | Yes |